Search
Search
| Gene Symbol: Aldob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012496.2 | NP_036628.2 | 8 - 9 | 1061 | 73 | |
| GenBank mRNA | BC081697.1 | - | 8 - 9 | 1061 | 7109764 | 73 |
| FQ209505.1 | - | 8 - 9 | 1082 | 73 | ||
| FQ209538.1 | - | 8 - 9 | 1080 | 73 | ||
| FQ209583.1 | - | 8 - 9 | 1082 | 73 | ||
| FQ209594.1 | - | 8 - 9 | 1080 | 73 | ||
| FQ209615.1 | - | 8 - 9 | 1080 | 73 | ||
| FQ209621.1 | - | 8 - 9 | 1080 | 73 | ||
| FQ209639.1 | - | 8 - 9 | 1082 | 73 | ||
| FQ209643.1 | - | 8 - 9 | 1078 | 73 | ||
| FQ209782.1 | - | 8 - 9 | 1083 | 73 | ||
| FQ209819.1 | - | 8 - 9 | 1091 | 73 | ||
| FQ210002.1 | - | 8 - 9 | 1083 | 73 | ||
| FQ210453.1 | - | 6 - 7 | 788 | 73 | ||
| FQ210849.1 | - | 8 - 9 | 1083 | 73 | ||
| FQ211083.1 | - | 8 - 9 | 1085 | 73 | ||
| FQ211156.1 | - | 8 - 9 | 1083 | 73 | ||
| FQ211159.1 | - | 8 - 9 | 1081 | 73 | ||
| FQ211293.1 | - | 6 - 7 | 759 | 73 | ||
| FQ218139.1 | - | 8 - 9 | 1082 | 73 | ||
| FQ218217.1 | - | 8 - 9 | 1082 | 73 | ||
| FQ218348.1 | - | 8 - 9 | 1088 | 73 | ||
| FQ218402.1 | - | 8 - 9 | 1084 | 73 | ||
| FQ218522.1 | - | 8 - 9 | 1081 | 73 | ||
| FQ218551.1 | - | 8 - 9 | 1084 | 73 | ||
| FQ218592.1 | - | 4 - 5 | 580 | 73 | ||
| FQ218665.1 | - | 8 - 9 | 1080 | 73 | ||
| FQ218911.1 | - | 8 - 9 | 1081 | 73 | ||
| FQ219077.1 | - | 6 - 7 | 765 | 73 | ||
| FQ219096.1 | - | 8 - 9 | 1082 | 73 | ||
| FQ219116.1 | - | 8 - 9 | 73 | |||
| FQ219236.1 | - | 8 - 9 | 1080 | 73 | ||
| FQ219261.1 | - | 8 - 9 | 1082 | 73 | ||
| FQ219453.1 | - | 8 - 9 | 1079 | 73 | ||
| FQ219574.1 | - | 8 - 9 | 1081 | 73 | ||
| FQ219585.1 | - | 8 - 9 | 1082 | 73 | ||
| FQ219640.1 | - | 8 - 9 | 1079 | 73 | ||
| FQ219646.1 | - | 8 - 9 | 1080 | 73 | ||
| FQ219703.1 | - | 8 - 9 | 1083 | 73 | ||
Set Membership: |
|