Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01768517_m1
          See other SLC25A16 GE Assays ›
          Gene Symbol:
          Slc25a16
          Gene Name:
          solute carrier family 25 (mitochondrial carrier), member 16
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 27283383 - 27308069 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01768517_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc25a16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100860.1 NP_001094330.1 1 - 2 198 58
          GenBank mRNA BC166494.1 - 1 - 2 198 7110872 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001486.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          mitochondrial carrier protein

          Gene Ontology Categories:

          Function(s)

          transmembrane transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P113999 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P542773 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P123695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18198 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114944 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10308 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20060 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P542773 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123695 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P113999 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123695 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P113999 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P542773 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P113999 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P542773 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P123695 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P461331 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68223 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10829838 - -
          Affymetrix Rat Gene 1.0 ST Array 10829832 - -
          Affymetrix Rat Gene 1.0 ST Array 10829835 - -
          Affymetrix Rat Gene 1.0 ST Array 10829834 - -
          Affymetrix Rat Gene 1.0 ST Array 10829833 - -
          Affymetrix Rat Gene 1.0 ST Array 10829836 - -
          Affymetrix Rat Gene 1.0 ST Array 10829839 - -
          Affymetrix Rat Gene 1.0 ST Array 10829837 - -
          Affymetrix Rat Gene 1.1 ST Array 10829831 - -
          Rat Genome 230 2.0 Array 1384368_at - -
          Rat Genome 230 2.0 Array 1384368_PM_at - -
          Affymetrix HT Rat Focus Array 1384368_PM_at - -
          Affymetrix HT Rat Focus Array 1384368_at - -
          Affymetrix Rat Clariom S Assay PSR2000012169.rn.1 TC2000001486.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000012171.rn.1 TC2000001486.rn.1 RTA TC
          Rat Expression Set 230 1384368_PM_at - -
          Rat Expression Set 230 1384368_at - -
          Affymetrix Rat Gene 1.0 ST Array 10829831 - -
          Rat Genome U34 Set rc_AA900084_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21447465 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366213 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline