Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01768654_m1
          See other RNF8 GE Assays ›
          Gene Symbol:
          Rnf8
          Gene Name:
          ring finger protein 8, E3 ubiquitin protein ligase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 8285338 - 8309858 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01768654_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025727.1 NP_001020898.1 2 - 3 321 66
          XM_006256225.2 XP_006256287.1 2 - 3 398 66
          XM_006256226.2 XP_006256288.1 2 - 3 401 66
          GenBank mRNA BC099079.1 - 2 - 3 321 7443362 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000322.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf8

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair
          double-strand break repair via nonhomologous end joining
          ubiquitin-dependent protein catabolic process
          cellular response to DNA damage stimulus
          mitotic nuclear division
          spermatid development
          biological_process
          response to ionizing radiation
          histone H2A ubiquitination
          histone H2B ubiquitination
          negative regulation of transcription elongation from RNA polymerase II promoter
          histone exchange
          isotype switching
          positive regulation of DNA repair
          cell division
          protein autoubiquitination
          protein K63-linked ubiquitination
          histone H2A K63-linked ubiquitination
          protein K48-linked ubiquitination
          molecular_function
          chromatin binding
          ubiquitin-protein transferase activity
          zinc ion binding
          acid-amino acid ligase activity
          ubiquitin protein ligase binding
          histone binding
          protein homodimerization activity
          ubiquitin protein ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P092906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197678 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31136 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131775 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26817 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092906 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092906 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P092906 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P993934 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70986 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78864 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10828910 - -
          Affymetrix Rat Gene 1.0 ST Array 10828907 - -
          Affymetrix Rat Gene 1.0 ST Array 10828908 - -
          Affymetrix Rat Gene 1.0 ST Array 10828909 - -
          Affymetrix Rat Gene 1.0 ST Array 10828906 - -
          Affymetrix Rat Gene 1.0 ST Array 10828905 - -
          Affymetrix Rat Gene 1.0 ST Array 10828912 - -
          Affymetrix Rat Gene 1.0 ST Array 10828911 - -
          Affymetrix Rat Gene 1.1 ST Array 10828904 - -
          Rat Genome 230 2.0 Array 1389069_PM_at - -
          Rat Genome 230 2.0 Array 1389069_at - -
          Affymetrix HT Rat Focus Array 1389069_PM_at - -
          Affymetrix HT Rat Focus Array 1389069_at - -
          Affymetrix Rat Clariom S Assay PSR2000003000.rn.1 TC2000000322.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000003001.rn.1 TC2000000322.rn.1 RTA TC
          Rat Expression Set 230 1389069_PM_at - -
          Rat Expression Set 230 1389069_at - -
          Affymetrix Rat Gene 1.0 ST Array 10828904 - -
          Rat Genome U34 Set rc_AA996944_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21985339 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375115 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline