Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01768852_m1
          See other L2HGDH GE Assays ›
          Gene Symbol:
          L2hgdh
          Gene Name:
          L-2-hydroxyglutarate dehydrogenase
          Gene Aliases:
          RGD1306250
          Chromosome Location:
          Chr.6: 92016560 - 92057643 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01768852_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: L2hgdh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108028.1 NP_001101498.1 1 - 2 159 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600002607.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular protein metabolic process
          oxidation-reduction process
          2-hydroxyglutarate dehydrogenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P459702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249196 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99636 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147395 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19290 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38887 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142541 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P459702 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P854615 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P459702 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P854615 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P459702 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P854615 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P459702 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97659 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73490 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17256 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10890352 - -
          Affymetrix Rat Gene 1.0 ST Array 10890349 - -
          Affymetrix Rat Gene 1.0 ST Array 10890344 - -
          Affymetrix Rat Gene 1.0 ST Array 10890348 - -
          Affymetrix Rat Gene 1.0 ST Array 10890345 - -
          Affymetrix Rat Gene 1.0 ST Array 10890351 - -
          Affymetrix Rat Gene 1.0 ST Array 10890353 - -
          Affymetrix Rat Gene 1.0 ST Array 10890346 - -
          Affymetrix Rat Gene 1.0 ST Array 10890347 - -
          Affymetrix Rat Gene 1.0 ST Array 10890350 - -
          Affymetrix Rat Gene 1.1 ST Array 10890342 - -
          Rat Genome 230 2.0 Array 1382462_PM_at - -
          Rat Genome 230 2.0 Array 1382462_at - -
          Affymetrix HT Rat Focus Array 1382462_at - -
          Affymetrix HT Rat Focus Array 1382462_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600018445.rn.1 TC0600002607.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600018444.rn.1 TC0600002607.rn.1 RTA TC
          Rat Expression Set 230 1382462_at - -
          Rat Expression Set 230 1382462_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10890342 - -
          Rat Genome U34 Set rc_AI232564_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22035270 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352595 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline