Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01768907_g1
          See other NDUFAF7 GE Assays ›
          Gene Symbol:
          Ndufaf7
          Gene Name:
          NADH dehydrogenase (ubiquinone) complex I, assembly factor 7
          Gene Aliases:
          RGD1311578
          Chromosome Location:
          Chr.6: 1534594 - 1545831 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01768907_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufaf7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008318.1 NP_001008319.1 9 - 10 1109 85
          XM_006239627.2 XP_006239689.1 9 - 10 1216 85
          GenBank mRNA BC083810.1 - 9 - 10 1109 7110144 85

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ndufaf7

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial respiratory chain complex I assembly
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P340316 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P028551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862195 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63845 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170145 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P028551 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P340316 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P340316 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P028551 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P340316 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P028551 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P340316 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61243 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50671 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10882455 - -
          Affymetrix Rat Gene 1.0 ST Array 10791617 - -
          Affymetrix Rat Gene 1.0 ST Array 10791616 - -
          Affymetrix Rat Gene 1.0 ST Array 10791615 - -
          Affymetrix Rat Gene 1.0 ST Array 10882459 - -
          Affymetrix Rat Gene 1.0 ST Array 10882458 - -
          Affymetrix Rat Gene 1.0 ST Array 10882460 - -
          Affymetrix Rat Gene 1.0 ST Array 10882457 - -
          Affymetrix Rat Gene 1.0 ST Array 10882456 - -
          Affymetrix Rat Gene 1.1 ST Array 10791614 - -
          Affymetrix Rat Gene 1.1 ST Array 10882454 - -
          Rat Genome 230 2.0 Array 1393274_at - -
          Rat Genome 230 2.0 Array 1393274_PM_at - -
          Rat Genome 230 2.0 Array 1374082_at - -
          Rat Genome 230 2.0 Array 1374082_PM_at - -
          Affymetrix HT Rat Focus Array 1374082_PM_at - -
          Affymetrix HT Rat Focus Array 1374082_at - -
          Affymetrix HT Rat Focus Array 1393274_at - -
          Affymetrix HT Rat Focus Array 1393274_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600000129.rn.1 TC0600000017.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600000124.rn.1 TC0600000017.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600000127.rn.1 TC0600000017.rn.1 RTA TC
          Rat Expression Set 230 1374082_PM_at - -
          Rat Expression Set 230 1374082_at - -
          Rat Expression Set 230 1393274_PM_at - -
          Rat Expression Set 230 1393274_at - -
          Affymetrix Rat Gene 1.0 ST Array 10882454 - -
          Rat Genome U34 Set rc_AI170882_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21129094 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359207 - -
          RatRef-12 Expression BeadChip ILMN_58165 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline