Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01769134_m1
          See other TRIB2 GE Assays ›
          Gene Symbol:
          Trib2
          Gene Name:
          tribbles pseudokinase 2
          Gene Aliases:
          RGD1564451
          Chromosome Location:
          Chr.6: 41016722 - 41041026 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01769134_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trib2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108015.2 NP_001101485.1 1 - 2 40 70
          XM_006239905.2 XP_006239967.1 1 - 2 1629 70
          GenBank mRNA BC168939.1 - 1 - 2 40 7103466 70
          FQ210645.1 - 1 - 2 73 70
          FQ231260.1 - 1 - 2 687 70

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC168939.1, NM_001108015.2 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of interleukin-10 biosynthetic process
          negative regulation of fat cell differentiation
          regulation of catalytic activity
          protein kinase activity
          ATP binding
          transcription factor binding
          ubiquitin protein ligase binding
          ubiquitin-protein transferase regulator activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P160568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227303 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180575 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29515 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P160568 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P160568 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P160568 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P160568 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P267610 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70557 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10889264 - -
          Affymetrix Rat Gene 1.0 ST Array 10889265 - -
          Affymetrix Rat Gene 1.1 ST Array 10889263 - -
          Rat Genome 230 2.0 Array 1385124_PM_at - -
          Rat Genome 230 2.0 Array 1388821_PM_at - -
          Rat Genome 230 2.0 Array 1385124_at - -
          Rat Genome 230 2.0 Array 1388821_at - -
          Affymetrix HT Rat Focus Array 1385124_at - -
          Affymetrix HT Rat Focus Array 1385124_PM_at - -
          Affymetrix HT Rat Focus Array 1388821_at - -
          Affymetrix HT Rat Focus Array 1388821_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600015681.rn.1 TC0600002167.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600015684.rn.1 TC0600002167.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600015679.rn.1 TC0600002167.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600015682.rn.1 TC0600002167.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600015683.rn.1 TC0600002167.rn.1 RTA TC
          Rat Expression Set 230 1388821_at - -
          Rat Expression Set 230 1388821_PM_at - -
          Rat Expression Set 230 1385124_PM_at - -
          Rat Expression Set 230 1385124_at - -
          Affymetrix Rat Gene 1.0 ST Array 10889263 - -
          Rat Genome U34 Set rc_AI010430_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368194 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline