Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01769610_m1
          See other SRP54A GE Assays ›
          Gene Symbol:
          Srp54a
          Gene Name:
          signal recognition particle 54A
          Gene Aliases:
          Srp54
          Chromosome Location:
          Chr.6: 75996631 - 76035763 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01769610_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Srp54a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053871.1 NP_446323.1 15 - 16 1669 65
          XM_006240097.2 XP_006240159.1 14 - 15 1510 65
          GenBank mRNA BC079117.1 - 15 - 16 1669 7126297 65
          FQ211580.1 - 15 - 16 1680 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000788.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Srp54a

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          SRP-dependent cotranslational protein targeting to membrane
          biological_process
          metabolic process
          response to drug
          protein targeting to ER
          molecular_function
          GTPase activity
          GTP binding
          drug binding
          7S RNA binding
          GDP binding
          endoplasmic reticulum signal peptide binding
          ribonucleoprotein complex binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P528007 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P088612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599475 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51113 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67016 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103130 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53584 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50941 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088612 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P528007 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P528007 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088612 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P088612 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P528007 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P378315 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P528007 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26166 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31583 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35444 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10884625 - -
          Affymetrix Rat Gene 1.0 ST Array 10884627 - -
          Affymetrix Rat Gene 1.0 ST Array 10884626 - -
          Affymetrix Rat Gene 1.0 ST Array 10884632 - -
          Affymetrix Rat Gene 1.0 ST Array 10884624 - -
          Affymetrix Rat Gene 1.0 ST Array 10884630 - -
          Affymetrix Rat Gene 1.0 ST Array 10884638 - -
          Affymetrix Rat Gene 1.0 ST Array 10884641 - -
          Affymetrix Rat Gene 1.0 ST Array 10884633 - -
          Affymetrix Rat Gene 1.0 ST Array 10884634 - -
          Affymetrix Rat Gene 1.0 ST Array 10884639 - -
          Affymetrix Rat Gene 1.0 ST Array 10884637 - -
          Affymetrix Rat Gene 1.0 ST Array 10884640 - -
          Affymetrix Rat Gene 1.0 ST Array 10884631 - -
          Affymetrix Rat Gene 1.0 ST Array 10884636 - -
          Affymetrix Rat Gene 1.0 ST Array 10884629 - -
          Affymetrix Rat Gene 1.0 ST Array 10884628 - -
          Affymetrix Rat Gene 1.0 ST Array 10884635 - -
          Affymetrix Rat Gene 1.1 ST Array 10884623 - -
          Rat Genome 230 2.0 Array 1383285_at - -
          Rat Genome 230 2.0 Array 1383410_at - -
          Rat Genome 230 2.0 Array 1383285_PM_at - -
          Rat Genome 230 2.0 Array 1383410_PM_at - -
          Rat Genome 230 2.0 Array 1385680_PM_at - -
          Rat Genome 230 2.0 Array 1385680_at - -
          Affymetrix HT Rat Focus Array 1383410_PM_at - -
          Affymetrix HT Rat Focus Array 1385680_PM_at - -
          Affymetrix HT Rat Focus Array 1385680_at - -
          Affymetrix HT Rat Focus Array 1383410_at - -
          Affymetrix Rat Clariom S Assay PSR0600005258.rn.1 TC0600000788.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600005259.rn.1 TC0600000788.rn.1 RTA TC
          Rat Expression Set 230 1385680_PM_at - -
          Rat Expression Set 230 1385680_at - -
          Rat Expression Set 230 1383410_PM_at - -
          Rat Expression Set 230 1383410_at - -
          Affymetrix Rat Gene 1.0 ST Array 10884623 - -
          Rat Genome U34 Set rc_AA924196_at - -
          Rat Genome U34 Set H32369_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21387148 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371927 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline