Search
Search
| Gene Symbol: Bcap29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006980.1 | NP_001006981.1 | 4 - 5 | 504 | 83 | |
| XM_006239982.2 | XP_006240044.1 | 5 - 6 | 625 | 83 | ||
| GenBank mRNA | BC083576.1 | - | 4 - 5 | 504 | 7113186 | 83 |
Set Membership: |
|