Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: She | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001062249.5 | XP_001062249.1 | 2 - 3 | 918 | 70 | |
XM_008761244.1 | XP_008759466.1 | 2 - 3 | 918 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Probe spans exons |