Search
Search
| Gene Symbol: Kirrel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207606.1 | NP_997489.1 | 11 - 12 | 1565 | 58 | |
| XM_006232675.2 | XP_006232737.1 | 12 - 13 | 1903 | 58 | ||
| XM_006232676.2 | XP_006232738.1 | 12 - 13 | 1900 | 58 | ||
| XM_006232677.2 | XP_006232739.1 | 12 - 13 | 1855 | 58 | ||
| XM_006232678.2 | XP_006232740.1 | 11 - 12 | 1757 | 58 | ||
| GenBank mRNA | AY249056.1 | - | 11 - 12 | 1565 | 58 | |
| AY271309.1 | - | 10 - 11 | 1314 | 58 | ||
Set Membership: |
|