Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01769975_m1
          See other PID1 GE Assays ›
          Gene Symbol:
          Pid1
          Gene Name:
          phosphotyrosine interaction domain containing 1
          Gene Aliases:
          Ab1-119, RGD1560766
          Chromosome Location:
          Chr.9: 91667748 - 91683468 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01769975_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pid1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110493.1 NP_001103963.1 3 - 4 299 65
          GenBank mRNA AY325140.1 - 3 - 4 299 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002365.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s)

          negative regulation of protein phosphorylation
          positive regulation of gene expression
          cellular response to leptin stimulus
          fat cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of glucose import
          negative regulation of insulin receptor signaling pathway
          regulation of mitochondrial membrane potential
          positive regulation of fat cell proliferation
          mitochondrion morphogenesis
          cellular response to cytokine stimulus
          cellular response to interleukin-6
          cellular response to tumor necrosis factor
          cellular response to fatty acid
          negative regulation of establishment of protein localization to plasma membrane
          negative regulation of mitochondrial DNA replication
          regulation of G1/S transition of mitotic cell cycle
          regulation of reactive oxygen species metabolic process
          positive regulation of reactive oxygen species metabolic process
          negative regulation of ATP biosynthetic process
          positive regulation of ATP biosynthetic process
          negative regulation of glucose import in response to insulin stimulus

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P545635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162018 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108102 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29917 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1118 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P545635 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P545635 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P545635 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P545635 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P592613 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58461 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10929475 - -
          Affymetrix Rat Gene 1.0 ST Array 10929477 - -
          Affymetrix Rat Gene 1.0 ST Array 10929474 - -
          Affymetrix Rat Gene 1.0 ST Array 10929476 - -
          Affymetrix Rat Gene 1.1 ST Array 10929473 - -
          Affymetrix Rat Clariom S Assay PSR0900019378.rn.1 TC0900002365.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10929473 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351973 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline