Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01771026_m1
          See other RGD1559493 GE Assays ›
          Gene Symbol:
          RGD1559493
          Gene Name:
          similar to Hypothetical protein MGC58608
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 126196071 - 126221916 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01771026_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1559493
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109262.2 NP_001102732.1 9 - 10 1187 63
          GenBank mRNA BC160900.1 - 9 - 10 1187 7454065 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003287.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P160650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347818 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76690 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170707 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148679 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160650 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160650 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P160650 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7320 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10878459 - -
          Affymetrix Rat Gene 1.0 ST Array 10878461 - -
          Affymetrix Rat Gene 1.0 ST Array 10878460 - -
          Affymetrix Rat Gene 1.0 ST Array 10878465 - -
          Affymetrix Rat Gene 1.0 ST Array 10878466 - -
          Affymetrix Rat Gene 1.0 ST Array 10878462 - -
          Affymetrix Rat Gene 1.0 ST Array 10878464 - -
          Affymetrix Rat Gene 1.0 ST Array 10878463 - -
          Affymetrix Rat Gene 1.0 ST Array 10878458 - -
          Affymetrix Rat Gene 1.1 ST Array 10878457 - -
          Affymetrix Rat Clariom S Assay PSR0500023814.rn.1 TC0500003287.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023818.rn.1 TC0500003287.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023815.rn.1 TC0500003287.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500023816.rn.1 TC0500003287.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10878457 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21885127 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371016 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline