Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01771475_m1
          See other SRSF12 GE Assays ›
          Gene Symbol:
          Srsf12
          Gene Name:
          serine/arginine-rich splicing factor 12
          Gene Aliases:
          RGD1561162, Srrp, Srrp35
          Chromosome Location:
          Chr.5: 48456777 - 48484017 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01771475_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Srsf12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135711.1 NP_001129183.1 4 - 5 421 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000405.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          mRNA 5'-splice site recognition
          nucleotide binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P046990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792857 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95138 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89418 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046990 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046990 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P046990 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P152219 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10868170 - -
          Affymetrix Rat Gene 1.0 ST Array 10868168 - -
          Affymetrix Rat Gene 1.0 ST Array 10868169 - -
          Affymetrix Rat Gene 1.0 ST Array 10868167 - -
          Affymetrix Rat Gene 1.0 ST Array 10868166 - -
          Affymetrix Rat Gene 1.1 ST Array 10868165 - -
          Affymetrix Rat Clariom S Assay PSR0500002522.rn.1 TC0500000405.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500002528.rn.1 TC0500000405.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500002531.rn.1 TC0500000405.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500002523.rn.1 TC0500000405.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500002527.rn.1 TC0500000405.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500002530.rn.1 TC0500000405.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500002526.rn.1 TC0500000405.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500002529.rn.1 TC0500000405.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10868165 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362326 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline