Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01771830_m1
          See other CGM4 GE Assays ›
          Gene Symbol:
          Cgm4
          Gene Name:
          carcinoembryonic antigen gene family 4
          Gene Aliases:
          CEA4, CEA5, CGM4AA, Cea, Cear, Psg16
          Chromosome Location:
          Chr.1: 79030291 - 79043259 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01771830_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cgm4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012525.1 NP_036657.1 2 - 3 631 80
          GenBank mRNA BC097366.1 - 2 - 3 595 7443149 80
          CB804911.1 - 1 - 2 94 80
          CV118170.1 - 2 - 3 595 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000956.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cgm4

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787372 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67471 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10989 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10989 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10989 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P305556 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10989 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73802 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54932 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10704584 - -
          Affymetrix Rat Gene 1.0 ST Array 10704586 - -
          Affymetrix Rat Gene 1.0 ST Array 10704587 - -
          Affymetrix Rat Gene 1.0 ST Array 10704588 - -
          Affymetrix Rat Gene 1.0 ST Array 10704585 - -
          Affymetrix Rat Gene 1.1 ST Array 10704583 - -
          Rat Genome 230 2.0 Array 1389856_PM_at - -
          Rat Genome 230 2.0 Array 1389856_at - -
          Rat Genome 230 2.0 Array 1382690_at - -
          Rat Genome 230 2.0 Array 1382690_PM_at - -
          Affymetrix HT Rat Focus Array 1389856_PM_at - -
          Affymetrix HT Rat Focus Array 1382690_PM_at - -
          Affymetrix HT Rat Focus Array 1389856_at - -
          Affymetrix HT Rat Focus Array 1382690_at - -
          Affymetrix Rat Clariom S Assay PSR0100006606.rn.1 TC0100000956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006613.rn.1 TC0100000956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006605.rn.1 TC0100000956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006612.rn.1 TC0100000956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006610.rn.1 TC0100000956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006609.rn.1 TC0100000956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006607.rn.1 TC0100000956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006608.rn.1 TC0100000956.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006611.rn.1 TC0100000956.rn.1 RTA TC
          Rat Expression Set 230 1389856_at - -
          Rat Expression Set 230 1382690_PM_at - -
          Rat Expression Set 230 1389856_PM_at - -
          Rat Expression Set 230 1382690_at - -
          Affymetrix Rat Gene 1.0 ST Array 10704583 - -
          Rat Genome U34 Set M32475cds_g_at - -
          Rat Genome U34 Set M32475cds_at - -
          Rat Genome U34 Set rc_AI179387_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22226478 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363112 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline