Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01772203_m1
          See other LOC502176 GE Assays ›
          Gene Symbol:
          LOC502176
          Gene Name:
          hypothetical protein LOC502176
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 60729517 - 60767280 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01772203_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC502176
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025057.2 NP_001020228.2 3 - 4 297 83
          GenBank mRNA AY325157.1 - 3 - 4 297 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000681.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC502176

          Panther Classification:

          Molecular Function -

          ribosomal protein

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P114049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274035 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178901 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153766 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115886 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44671 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99650 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86131 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168945 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85883 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P114049 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P114049 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P114049 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P168631 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17800 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60425 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78218 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86866 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104497 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66692 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10802268 - -
          Affymetrix Rat Gene 1.0 ST Array 10802270 - -
          Affymetrix Rat Gene 1.0 ST Array 10802271 - -
          Affymetrix Rat Gene 1.0 ST Array 10802269 - -
          Affymetrix Rat Gene 1.1 ST Array 10802267 - -
          Affymetrix Rat Clariom S Assay PSR1800005009.rn.1 TC1800000681.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10802267 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366368 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0