Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01772426_m1
          See other VOM2R60 GE Assays ›
          Gene Symbol:
          Vom2r60
          Gene Name:
          vomeronasal 2 receptor, 60
          Gene Aliases:
          RGD1560561
          Chromosome Location:
          Chr.12: 4054395 - 4436352 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01772426_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vom2r60
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001099480.1 NP_001092950.1 6 - 7 1677 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000076.rn.1

          Set Membership:

          Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P186630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258046 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1276 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168332 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143567 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 374 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158816 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85438 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22833 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12626 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55199 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P186630 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P186630 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P186630 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P189629 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52949 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3892 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31480 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10756067 - -
          Affymetrix Rat Gene 1.0 ST Array 10756072 - -
          Affymetrix Rat Gene 1.0 ST Array 10756073 - -
          Affymetrix Rat Gene 1.0 ST Array 10756060 - -
          Affymetrix Rat Gene 1.0 ST Array 10756070 - -
          Affymetrix Rat Gene 1.0 ST Array 10756068 - -
          Affymetrix Rat Gene 1.0 ST Array 10756074 - -
          Affymetrix Rat Gene 1.0 ST Array 10756062 - -
          Affymetrix Rat Gene 1.0 ST Array 10756063 - -
          Affymetrix Rat Gene 1.0 ST Array 10756064 - -
          Affymetrix Rat Gene 1.0 ST Array 10756071 - -
          Affymetrix Rat Gene 1.1 ST Array 10756069 - -
          Affymetrix Rat Gene 1.1 ST Array 10756059 - -
          Affymetrix Rat Clariom S Assay PSR1200000653.rn.1 TC1200000076.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10756069 - -
          Affymetrix Rat Gene 1.0 ST Array 10756059 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21431243 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366049 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline