Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01772582_m1
          See other TCP10B GE Assays ›
          Gene Symbol:
          Tcp10b
          Gene Name:
          t-complex protein 10b
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 53531007 - 53554113 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01772582_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tcp10b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107461.1 NP_001100931.1 1 - 2 107 59
          XM_006227927.2 XP_006227989.1 3 - 4 377 59
          XM_006227928.2 XP_006227990.1 3 - 4 377 59
          XM_006227930.2 XP_006227992.1 3 - 4 377 59
          XM_008758757.1 XP_008756979.1 1 - 2 148 59
          XM_008758758.1 XP_008756980.1 1 - 2 148 59
          XM_008758759.1 XP_008756981.1 2 - 3 191 59
          GenBank mRNA BC166440.1 - 3 - 4 308 7455355 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000563.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          cytoskeletal protein microtubule or microtubule-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          RNA polymerase II transcription corepressor activity
          identical protein binding
          protein self-association
          repressing transcription factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P304432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721519 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126487 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P304432 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P304432 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P304432 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P304432 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65693 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10703181 - -
          Affymetrix Rat Gene 1.0 ST Array 10703182 - -
          Affymetrix Rat Gene 1.0 ST Array 10703175 - -
          Affymetrix Rat Gene 1.0 ST Array 10703173 - -
          Affymetrix Rat Gene 1.0 ST Array 10703171 - -
          Affymetrix Rat Gene 1.0 ST Array 10703179 - -
          Affymetrix Rat Gene 1.0 ST Array 10703177 - -
          Affymetrix Rat Gene 1.0 ST Array 10703172 - -
          Affymetrix Rat Gene 1.0 ST Array 10703176 - -
          Affymetrix Rat Gene 1.0 ST Array 10703180 - -
          Affymetrix Rat Gene 1.0 ST Array 10703174 - -
          Affymetrix Rat Gene 1.1 ST Array 10703169 - -
          Affymetrix Rat Clariom S Assay PSR0100003592.rn.1 TC0100000563.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100003591.rn.1 TC0100000563.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100003595.rn.1 TC0100000563.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100003593.rn.1 TC0100000563.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100003597.rn.1 TC0100000563.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100003598.rn.1 TC0100000563.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10703169 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22321151 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362031 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline