Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01772770_m1
          See other TMEM196 GE Assays ›
          Gene Symbol:
          Tmem196
          Gene Name:
          transmembrane protein 196
          Gene Aliases:
          RGD1562527
          Chromosome Location:
          Chr.6: 147875011 - 147940231 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01772770_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem196
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001044269.1 NP_001037734.1 3 - 4 975 66
          XM_006240702.2 XP_006240764.1 3 - 4 2219 66
          GenBank mRNA BC089997.1 - 3 - 4 975 7320460 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001718.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P852545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899541 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66689 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154118 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173526 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111800 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89701 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74531 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P852545 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P852545 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P852545 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P852545 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29726 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70295 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69637 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58666 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9198 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10887828 - -
          Affymetrix Rat Gene 1.0 ST Array 10887827 - -
          Affymetrix Rat Gene 1.0 ST Array 10887829 - -
          Affymetrix Rat Gene 1.0 ST Array 10887830 - -
          Affymetrix Rat Gene 1.1 ST Array 10887826 - -
          Affymetrix Rat Clariom S Assay PSR0600012134.rn.1 TC0600001718.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012136.rn.1 TC0600001718.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012133.rn.1 TC0600001718.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012135.rn.1 TC0600001718.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10887826 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21080196 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370620 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline