Search
Search

Mien1
LOC102552549| Gene Symbol: Mien1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108296.1 | NP_001101766.1 | 1 - 2 | 136 | 147 | |
| GenBank mRNA | FQ214212.1 | - | 133 | 147 | ||
| Gene Symbol: LOC102552549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247244.2 | XP_006247306.1 | 1 - 2 | 449 | 147 | |
Set Membership: |
|