Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01774279_m1
          See other ZFP503 GE Assays ›
          Gene Symbol:
          Zfp503
          Gene Name:
          zinc finger protein 503
          Gene Aliases:
          Nolz-1, Nolz1, Znf503
          Chromosome Location:
          Chr.15: 2374582 - 2378370 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01774279_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp503
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107250.1 NP_001100720.1 1 - 2 765 123
          GenBank mRNA BC166993.1 - 1 - 2 780 123

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000016.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cell proliferation
          negative regulation of gene expression
          neural precursor cell proliferation
          G1 to G0 transition involved in cell differentiation
          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P090117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327575 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P625157 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P625157 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P625157 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P447839 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P625157 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10778995 - -
          Affymetrix Rat Gene 1.0 ST Array 10778996 - -
          Affymetrix Rat Gene 1.1 ST Array 10778993 - -
          Rat Genome 230 2.0 Array 1377241_PM_at - -
          Rat Genome 230 2.0 Array 1386721_PM_at - -
          Rat Genome 230 2.0 Array 1375366_PM_at - -
          Rat Genome 230 2.0 Array 1375366_at - -
          Rat Genome 230 2.0 Array 1393990_at - -
          Rat Genome 230 2.0 Array 1386721_at - -
          Rat Genome 230 2.0 Array 1377241_at - -
          Rat Genome 230 2.0 Array 1393990_PM_at - -
          Affymetrix HT Rat Focus Array 1393990_at - -
          Affymetrix HT Rat Focus Array 1386721_PM_at - -
          Affymetrix HT Rat Focus Array 1377241_at - -
          Affymetrix HT Rat Focus Array 1377241_PM_at - -
          Affymetrix HT Rat Focus Array 1393990_PM_at - -
          Affymetrix HT Rat Focus Array 1386721_at - -
          Affymetrix Rat Clariom S Assay PSR1500000123.rn.1 TC1500000016.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500000124.rn.1 TC1500000016.rn.1 RTA TC
          Rat Expression Set 230 1377241_at - -
          Rat Expression Set 230 1377241_PM_at - -
          Rat Expression Set 230 1375366_PM_at - -
          Rat Expression Set 230 1375366_at - -
          Rat Expression Set 230 1386721_PM_at - -
          Rat Expression Set 230 1386721_at - -
          Rat Expression Set 230 1393990_at - -
          Rat Expression Set 230 1393990_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10778993 - -
          Rat Genome U34 Set rc_AI028967_at - -
          Rat Genome U34 Set rc_AA956230_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21184728 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367725 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline