Search
Search
| Gene Symbol: Gdf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170350.1 | NP_001163821.1 | 2 | 1179 | 113 | |
| XM_006239878.2 | XP_006239940.1 | 2 | 1284 | 113 | ||
| GenBank mRNA | AB087406.1 | - | 1 | 55 | 113 | |
Set Membership: |
|