Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01777275_sH
          See other VOM1R105 GE Assays ›
          Gene Symbol:
          Vom1r105
          Gene Name:
          vomeronasal 1 receptor 105
          Gene Aliases:
          Vmn1r51, Vn1, Vnr1
          Chromosome Location:
          Chr.4: 122124451 - 122131893 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01777275_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vom1r105
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173113.1 NP_775136.1 2 1267 134
          GenBank mRNA U36785.1 - 2 1267 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001393.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vom1r105

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P034966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429646 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6499 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15235 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29028 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115653 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147819 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132472 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146720 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21205 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35760 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115938 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73078 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110502 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124807 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129783 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73036 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P200905 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98025 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11419 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27170 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12428 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44904 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101249 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84982 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49959 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10864047 - -
          Affymetrix Rat Gene 1.0 ST Array 10857214 - -
          Affymetrix Rat Gene 1.0 ST Array 10864044 - -
          Affymetrix Rat Gene 1.0 ST Array 10864043 - -
          Affymetrix Rat Gene 1.0 ST Array 10864045 - -
          Affymetrix Rat Gene 1.0 ST Array 10857213 - -
          Affymetrix Rat Gene 1.1 ST Array 10857212 - -
          Affymetrix Rat Gene 1.1 ST Array 10864042 - -
          Rat Genome 230 2.0 Array 1370726_at - -
          Rat Genome 230 2.0 Array 1370726_PM_at - -
          Affymetrix HT Rat Focus Array 1370726_at - -
          Affymetrix HT Rat Focus Array 1370726_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400009871.rn.1 TC0400001393.rn.1 RTA TC
          Rat Expression Set 230 1370726_PM_at - -
          Rat Expression Set 230 1370726_at - -
          Affymetrix Rat Gene 1.0 ST Array 10857212 - -
          Affymetrix Rat Gene 1.0 ST Array 10864042 - -
          Rat Genome U34 Set U36785_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20701829 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360751 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline