Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001000342.1 | NP_001000342.1 | 55 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus Inventoried |