Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01784385_g1
          See other OLR1366 GE Assays ›
          Gene Symbol:
          Olr1366
          Gene Name:
          olfactory receptor 1366
          Gene Aliases:
          tpcr13
          Chromosome Location:
          Chr.10: 12466153 - 12477674 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01784385_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olr1366
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001000980.1 NP_001000980.1 1 906 80
          XM_008767521.1 XP_008765743.1 2 1056 80
          XM_008767522.1 XP_008765744.1 2 995 80
          XM_008767523.1 XP_008765745.1 5 1077 80
          GenBank mRNA BC097272.1 - 5 1047 80

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olr1366

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P074665 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P16375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484925 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171744 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58363 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16375 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P074665 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P074665 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16375 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P16375 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P074665 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16375 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81068 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10740786 - -
          Affymetrix Rat Gene 1.0 ST Array 10749817 - -
          Affymetrix Rat Gene 1.0 ST Array 10740788 - -
          Affymetrix Rat Gene 1.0 ST Array 10740785 - -
          Affymetrix Rat Gene 1.0 ST Array 10740805 - -
          Affymetrix Rat Gene 1.0 ST Array 10740787 - -
          Affymetrix Rat Gene 1.0 ST Array 10740712 - -
          Affymetrix Rat Gene 1.0 ST Array 10740789 - -
          Affymetrix Rat Gene 1.1 ST Array 10740784 - -
          Affymetrix Rat Clariom S Assay PSR1000020660.rn.1 TC1000002333.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000020661.rn.1 TC1000002333.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000020662.rn.1 TC1000002333.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000020663.rn.1 TC1000002333.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10740711 - -
          Affymetrix Rat Gene 1.0 ST Array 10740784 - -
          Affymetrix Rat Gene 1.0 ST Array 10749816 - -
          Affymetrix Rat Gene 1.0 ST Array 10740804 - -
          Rat Genome U34 Set X89701cds_i_at - -
          Rat Genome U34 Set X89701cds_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22123540 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364974 - -
          RatRef-12 Expression BeadChip ILMN_59339 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline