Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01786289_m1
          See other ST3GAL4 GE Assays ›
          Gene Symbol:
          St3gal4
          Gene Name:
          ST3 beta-galactoside alpha-2,3-sialyltransferase 4
          Gene Aliases:
          siat4c
          Chromosome Location:
          Chr.8: 36264741 - 36314826 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01786289_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: St3gal4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_203337.2 NP_976082.1 9 - 10 1007 73
          XM_006242789.2 XP_006242851.1 10 - 11 1123 73
          XM_008766058.1 XP_008764280.1 9 - 10 1509 73
          XM_008766059.1 XP_008764281.1 9 - 10 1498 73
          XM_008766060.1 XP_008764282.1 9 - 10 1957 73
          GenBank mRNA AJ626825.1 - 8 - 9 771 73
          BC089057.1 - 9 - 10 1007 7315905 73
          FQ226042.1 - 2 - 3 252 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002209.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene St3gal4

          Panther Classification:

          Molecular Function -

          transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          cognition
          sialylation
          beta-galactoside (CMP) alpha-2,3-sialyltransferase activity
          protein binding
          monosialoganglioside sialyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P740939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934465 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57588 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38258 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100747 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26887 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137077 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P740939 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P740939 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P740939 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P461187 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15044 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87067 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10729 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57299 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86286 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10916025 - -
          Affymetrix Rat Gene 1.0 ST Array 10916017 - -
          Affymetrix Rat Gene 1.0 ST Array 10916027 - -
          Affymetrix Rat Gene 1.0 ST Array 10916019 - -
          Affymetrix Rat Gene 1.0 ST Array 10916024 - -
          Affymetrix Rat Gene 1.0 ST Array 10916023 - -
          Affymetrix Rat Gene 1.0 ST Array 10916021 - -
          Affymetrix Rat Gene 1.0 ST Array 10916018 - -
          Affymetrix Rat Gene 1.0 ST Array 10916026 - -
          Affymetrix Rat Gene 1.0 ST Array 10916022 - -
          Affymetrix Rat Gene 1.0 ST Array 10916020 - -
          Affymetrix Rat Gene 1.1 ST Array 10916016 - -
          Rat Genome 230 2.0 Array 1392881_at - -
          Rat Genome 230 2.0 Array 1392881_PM_at - -
          Rat Genome 230 2.0 Array 1373224_PM_at - -
          Rat Genome 230 2.0 Array 1373224_at - -
          Affymetrix HT Rat Focus Array 1392881_PM_at - -
          Affymetrix HT Rat Focus Array 1373224_PM_at - -
          Affymetrix HT Rat Focus Array 1392881_at - -
          Affymetrix HT Rat Focus Array 1373224_at - -
          Affymetrix Rat Clariom S Assay PSR0800017747.rn.1 TC0800002209.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017748.rn.1 TC0800002209.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017743.rn.1 TC0800002209.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017742.rn.1 TC0800002209.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017745.rn.1 TC0800002209.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017746.rn.1 TC0800002209.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017744.rn.1 TC0800002209.rn.1 RTA TC
          Rat Expression Set 230 1373224_PM_at - -
          Rat Expression Set 230 1373224_at - -
          Rat Expression Set 230 1392881_PM_at - -
          Rat Expression Set 230 1392881_at - -
          Affymetrix Rat Gene 1.0 ST Array 10916016 - -
          Rat Genome U34 Set rc_AI237091_at - -
          Rat Genome U34 Set rc_AI176688_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21997430 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361562 - -
          RatRef-12 Expression BeadChip ILMN_57996 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline