Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01789812_g1
          See other UBC GE Assays ›
          Gene Symbol:
          Ubc
          Gene Name:
          ubiquitin C
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 36638457 - 36642734 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01789812_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ubc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017314.1 NP_059010.1 2465 88
          GenBank mRNA BC089218.1 - 1 1971 88
          BC103477.1 - 3 1350 7929586 88
          D17296.1 - 2465 88
          FQ212515.1 - 1 988 88
          FQ229492.1 - 1 933 88
          X92660.1 - 1 56 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Endogenous Controls Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Chemokine Panel Rat Endogenous Controls 1700 Inventoried Rat_Endogenous_Controls Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ubc

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          ubiquitin homeostasis
          protease binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P048776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668027 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P048776 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P048776 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P048776 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1019957 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P109638 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10758136 - -
          Affymetrix Rat Gene 1.0 ST Array 10758135 - -
          Affymetrix Rat Gene 1.1 ST Array 10758134 - -
          Rat Genome 230 2.0 Array 1398767_PM_at - -
          Rat Genome 230 2.0 Array 1398767_at - -
          Affymetrix HT Rat Focus Array 1398767_at - -
          Affymetrix HT Rat Focus Array 1398767_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200004842.rn.1 TC1200000577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200004849.rn.1 TC1200000577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200004840.rn.1 TC1200000577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200004850.rn.1 TC1200000577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200004851.rn.1 TC1200000577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200004844.rn.1 TC1200000577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200004845.rn.1 TC1200000577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200004848.rn.1 TC1200000577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200004841.rn.1 TC1200000577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200004846.rn.1 TC1200000577.rn.1 RTA TC
          Rat Expression Set 230 1398767_PM_at - -
          Rat Expression Set 230 1398767_at - -
          Affymetrix Rat Gene 1.0 ST Array 10758134 - -
          Rat Genome U34 Set D17296_at - -
          Rat Genome U34 Set rc_AA819737_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22415427 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350494 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline