Search
Search
| Gene Symbol: Gjb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053388.1 | NP_445840.1 | 3 | 1663 | 142 | |
| XM_006252079.2 | XP_006252141.1 | 3 | 2989 | 142 | ||
| XM_006252080.2 | XP_006252142.1 | 3 | 2565 | 142 | ||
| XM_008770781.1 | XP_008769003.1 | 3 | 1777 | 142 | ||
| XM_008770782.1 | XP_008769004.1 | 3 | 2087 | 142 | ||
| XM_008770783.1 | XP_008769005.1 | 3 | 1668 | 142 | ||
| XM_008770784.1 | XP_008769006.1 | 3 | 1774 | 142 | ||
| GenBank mRNA | AF170284.1 | - | 3 | 1663 | 142 | |
| BC078753.1 | - | 3 | 1660 | 7132296 | 142 | |
Set Membership: |
|