Search
Search
| Gene Symbol: Mageb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106946.1 | NP_001100416.1 | 2 | 1172 | 143 | |
| XM_006257012.2 | XP_006257074.1 | 2 | 1146 | 143 | ||
| XM_006257013.2 | XP_006257075.1 | 2 | 1150 | 143 | ||
Set Membership: |
|