Search
Search

RGD1562726| Gene Symbol: RGD1562726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109052.1 | NP_001102522.1 | 3 | 2382 | 142 | |
| XM_006248074.2 | XP_006248136.1 | 3 | 2685 | 142 | ||
| XM_006248075.2 | XP_006248137.1 | 2 | 2115 | 142 | ||
| XM_006248076.2 | XP_006248138.1 | 3 | 2198 | 142 | ||
| XM_008768588.1 | XP_008766810.1 | 3 | 2664 | 142 | ||
| XM_008768589.1 | XP_008766811.1 | 2 | 2094 | 142 | ||
| XM_008768590.1 | XP_008766812.1 | 4 | 3719 | 142 | ||
| XM_008768591.1 | XP_008766813.1 | 3 | 2177 | 142 | ||
| XM_008768592.1 | XP_008766814.1 | 3 | 3136 | 142 | ||
| XM_008768593.1 | XP_008766815.1 | 3 | 2266 | 142 | ||
| XM_008768594.1 | XP_008766816.1 | 4 | 2870 | 142 | ||
| XM_008768595.1 | XP_008766817.1 | 4 | 2826 | 142 | ||
| XM_008768596.1 | XP_008766818.1 | 2 | 2453 | 142 | ||
| XM_008768597.1 | XP_008766819.1 | 2 | 2227 | 142 | ||
Set Membership: |
|