Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02094913_s1
          See other KCNE2 GE Assays ›
          Gene Symbol:
          Kcne2
          Gene Name:
          potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 2
          Gene Aliases:
          Mirp1
          Chromosome Location:
          Chr.11: 32434786 - 32447264 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02094913_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcne2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133603.2 NP_598287.1 2 1498 156
          XM_006248023.2 XP_006248085.1 2 1924 156
          XM_008768554.1 XP_008766776.1 2 1734 156
          GenBank mRNA BC089778.1 - 2 1498 7301610 156

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcne2

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          aging
          potassium ion import
          cellular protein localization
          cellular response to drug
          tongue development
          regulation of membrane repolarization
          regulation of ventricular cardiac muscle cell membrane repolarization
          potassium ion export
          potassium ion transmembrane transport
          cardiac muscle cell action potential involved in contraction
          ventricular cardiac muscle cell action potential
          membrane repolarization
          membrane repolarization during action potential
          regulation of heart rate by cardiac conduction
          regulation of potassium ion transmembrane transport
          positive regulation of voltage-gated calcium channel activity
          positive regulation of proteasomal protein catabolic process
          regulation of inward rectifier potassium channel activity
          regulation of cyclic nucleotide-gated ion channel activity
          regulation of delayed rectifier potassium channel activity
          negative regulation of voltage-gated potassium channel activity
          inward rectifier potassium channel activity
          voltage-gated potassium channel activity
          delayed rectifier potassium channel activity
          cation channel activity
          protein binding
          potassium channel regulator activity
          protein homodimerization activity
          ion channel binding
          voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P072973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948714 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139599 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072973 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072973 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P072973 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P414582 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80653 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10750288 - -
          Affymetrix Rat Gene 1.0 ST Array 10750289 - -
          Affymetrix Rat Gene 1.0 ST Array 10750287 - -
          Affymetrix Rat Gene 1.1 ST Array 10750286 - -
          Rat Genome 230 2.0 Array 1395150_at - -
          Rat Genome 230 2.0 Array 1369322_at - -
          Rat Genome 230 2.0 Array 1394343_s_at - -
          Rat Genome 230 2.0 Array 1394343_PM_s_at - -
          Rat Genome 230 2.0 Array 1386770_x_at - -
          Rat Genome 230 2.0 Array 1395150_PM_at - -
          Rat Genome 230 2.0 Array 1386770_PM_x_at - -
          Rat Genome 230 2.0 Array 1369322_PM_at - -
          Affymetrix HT Rat Focus Array 1395150_at - -
          Affymetrix HT Rat Focus Array 1369322_PM_at - -
          Affymetrix HT Rat Focus Array 1386770_PM_x_at - -
          Affymetrix HT Rat Focus Array 1395150_PM_at - -
          Affymetrix HT Rat Focus Array 1369322_at - -
          Affymetrix HT Rat Focus Array 1386770_x_at - -
          Rat Expression Set 230 1369322_PM_at - -
          Rat Expression Set 230 1394343_s_at - -
          Rat Expression Set 230 1386770_x_at - -
          Rat Expression Set 230 1369322_at - -
          Rat Expression Set 230 1394343_PM_s_at - -
          Rat Expression Set 230 1386770_PM_x_at - -
          Rat Expression Set 230 1395150_at - -
          Rat Expression Set 230 1395150_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10750286 - -
          Rat Genome U34 Set rc_AA850474_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21058910 - -
          ABI Rat Genome Survey Microarray 21728916 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651037 - -
          RatRef-12 Expression BeadChip ILMN_59878 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0