Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02102637_s1
          See other GTF3C6 GE Assays ›
          Gene Symbol:
          Gtf3c6
          Gene Name:
          general transcription factor IIIC, polypeptide 6, alpha
          Gene Aliases:
          RGD1307434
          Chromosome Location:
          Chr.20: 45347788 - 45356868 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02102637_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gtf3c6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108537.1 NP_001102007.1 6 1492 95
          GenBank mRNA BC091233.1 - 977 95
          BC159437.1 - 982 9102105 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          transcription from RNA polymerase III promoter
          biological_process
          molecular_function
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P095505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108767 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62496 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095504 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095504 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P095504 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P473472 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10833653 - -
          Affymetrix Rat Gene 1.0 ST Array 10833657 - -
          Affymetrix Rat Gene 1.0 ST Array 10833658 - -
          Affymetrix Rat Gene 1.0 ST Array 10833655 - -
          Affymetrix Rat Gene 1.0 ST Array 10833656 - -
          Affymetrix Rat Gene 1.0 ST Array 10833654 - -
          Affymetrix Rat Gene 1.1 ST Array 10833652 - -
          Rat Genome 230 2.0 Array 1374848_at - -
          Rat Genome 230 2.0 Array 1374848_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000015125.rn.1 TC2000001831.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000015118.rn.1 TC2000001831.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000015128.rn.1 TC2000001831.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000015126.rn.1 TC2000001831.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000015130.rn.1 TC2000001831.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000015119.rn.1 TC2000001831.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000015120.rn.1 TC2000001831.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000015124.rn.1 TC2000001831.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000015122.rn.1 TC2000001831.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000015127.rn.1 TC2000001831.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000015121.rn.1 TC2000001831.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000015129.rn.1 TC2000001831.rn.1 RTA TC
          Rat Expression Set 230 1374848_PM_at - -
          Rat Expression Set 230 1374848_at - -
          Affymetrix Rat Gene 1.0 ST Array 10833652 - -
          Rat Genome U34 Set rc_AA955913_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21680461 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364751 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline