Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02108275_s1
          See other TPST2 GE Assays ›
          Gene Symbol:
          Tpst2
          Gene Name:
          tyrosylprotein sulfotransferase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 50343076 - 50385098 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02108275_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tpst2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008508.2 NP_001008508.1 2 801 155
          XM_006249526.2 XP_006249588.1 3 881 155
          XM_008769319.1 XP_008767541.1 3 815 155
          GenBank mRNA BC086518.1 - 2 806 7302513 155

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tpst2

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-tyrosine sulfation
          fusion of sperm to egg plasma membrane
          prevention of polyspermy
          protein-tyrosine sulfotransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P206244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405170 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71676 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10921 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P206244 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P206244 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P206244 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P206244 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7390 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70274 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806156 - -
          Affymetrix Rat Gene 1.0 ST Array 10806153 - -
          Affymetrix Rat Gene 1.0 ST Array 10806154 - -
          Affymetrix Rat Gene 1.0 ST Array 10806155 - -
          Affymetrix Rat Gene 1.0 ST Array 10806152 - -
          Affymetrix Rat Gene 1.0 ST Array 10806157 - -
          Affymetrix Rat Gene 1.1 ST Array 10806151 - -
          Rat Genome 230 2.0 Array 1398994_at - -
          Rat Genome 230 2.0 Array 1398994_PM_at - -
          Affymetrix HT Rat Focus Array 1398994_at - -
          Affymetrix HT Rat Focus Array 1398994_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200015629.rn.1 TC1200001827.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015630.rn.1 TC1200001827.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015634.rn.1 TC1200001827.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015631.rn.1 TC1200001827.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015625.rn.1 TC1200001827.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015633.rn.1 TC1200001827.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015626.rn.1 TC1200001827.rn.1 RTA TC
          Rat Expression Set 230 1398994_at - -
          Rat Expression Set 230 1398994_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10806151 - -
          Rat Genome U34 Set rc_AI230778_at - -
          Rat Genome U34 Set rc_AI230778_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22134408 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362370 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530383 - -
          RatRef-12 Expression BeadChip ILMN_53520 - -
          RatRef-12 Expression BeadChip ILMN_161556 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline