Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02109307_u1
          See other PPA1 GE Assays ›
          Gene Symbol:
          Ppa1
          Gene Name:
          pyrophosphatase (inorganic) 1
          Gene Aliases:
          Pp, Pyp
          Chromosome Location:
          Chr.20: 31265519 - 31292451 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02109307_u1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100834.1 NP_001094304.1 2 240 142
          GenBank mRNA BC099794.1 - 2 240 142
          FQ215196.1 - 2 192 142
          FQ215675.1 - 2 192 142

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000685.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Amplicon spans exons and probe does not span exons

          Panther Classification:

          Molecular Function -

          pyrophosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphate-containing compound metabolic process
          magnesium ion binding
          inorganic diphosphatase activity
          pyrophosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P548520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118216 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50527 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17756 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119596 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P548520 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P548520 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P548520 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P548520 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27736 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73304 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10830045 - -
          Affymetrix Rat Gene 1.0 ST Array 10830048 - -
          Affymetrix Rat Gene 1.0 ST Array 10830043 - -
          Affymetrix Rat Gene 1.0 ST Array 10830044 - -
          Affymetrix Rat Gene 1.0 ST Array 10830041 - -
          Affymetrix Rat Gene 1.0 ST Array 10830046 - -
          Affymetrix Rat Gene 1.0 ST Array 10830042 - -
          Affymetrix Rat Gene 1.0 ST Array 10830047 - -
          Affymetrix Rat Gene 1.0 ST Array 10830040 - -
          Affymetrix Rat Gene 1.0 ST Array 10830039 - -
          Affymetrix Rat Gene 1.0 ST Array 10830049 - -
          Affymetrix Rat Gene 1.1 ST Array 10830038 - -
          Rat Genome 230 2.0 Array 1388960_PM_at - -
          Rat Genome 230 2.0 Array 1388960_at - -
          Affymetrix HT Rat Focus Array 1388960_at - -
          Affymetrix HT Rat Focus Array 1388960_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000006002.rn.1 TC2000000685.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006006.rn.1 TC2000000685.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006011.rn.1 TC2000000685.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006008.rn.1 TC2000000685.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006013.rn.1 TC2000000685.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006009.rn.1 TC2000000685.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006014.rn.1 TC2000000685.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006003.rn.1 TC2000000685.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006005.rn.1 TC2000000685.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006012.rn.1 TC2000000685.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006007.rn.1 TC2000000685.rn.1 RTA TC
          Rat Expression Set 230 1388960_at - -
          Rat Expression Set 230 1388960_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10830038 - -
          Rat Genome U34 Set rc_AA965166_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21876191 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370202 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline