Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02109317_s1
          See other SERINC1 GE Assays ›
          Gene Symbol:
          Serinc1
          Gene Name:
          serine incorporator 1
          Gene Aliases:
          LRRGT00191, Tde1l, Tde2
          Chromosome Location:
          Chr.20: 38966016 - 38985056 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02109317_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serinc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_182951.1 NP_891996.1 10 2183 146
          GenBank mRNA AY339894.1 - 10 2183 146
          BC088852.1 - 10 2212 7315523 146
          FQ212594.1 - 1 835 146
          FQ213445.1 - 1 490 146
          FQ214185.1 - 1 669 146

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serinc1

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidylserine metabolic process
          sphingolipid metabolic process
          phospholipid biosynthetic process
          L-serine transport
          positive regulation of transferase activity
          L-serine transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P321174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517199 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6047 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P321174 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P321174 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P321174 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P306325 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32589 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97276 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23458 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82577 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10833458 - -
          Affymetrix Rat Gene 1.0 ST Array 10833453 - -
          Affymetrix Rat Gene 1.0 ST Array 10833455 - -
          Affymetrix Rat Gene 1.0 ST Array 10833462 - -
          Affymetrix Rat Gene 1.0 ST Array 10833457 - -
          Affymetrix Rat Gene 1.0 ST Array 10833454 - -
          Affymetrix Rat Gene 1.0 ST Array 10833463 - -
          Affymetrix Rat Gene 1.0 ST Array 10833452 - -
          Affymetrix Rat Gene 1.0 ST Array 10833460 - -
          Affymetrix Rat Gene 1.0 ST Array 10833459 - -
          Affymetrix Rat Gene 1.0 ST Array 10833456 - -
          Affymetrix Rat Gene 1.1 ST Array 10833451 - -
          Rat Genome 230 2.0 Array 1388307_PM_at - -
          Rat Genome 230 2.0 Array 1388307_at - -
          Affymetrix HT Rat Focus Array 1388307_at - -
          Affymetrix HT Rat Focus Array 1388307_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000013369.rn.1 TC2000001625.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013370.rn.1 TC2000001625.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013368.rn.1 TC2000001625.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013365.rn.1 TC2000001625.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013362.rn.1 TC2000001625.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013366.rn.1 TC2000001625.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013360.rn.1 TC2000001625.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013372.rn.1 TC2000001625.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013367.rn.1 TC2000001625.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013361.rn.1 TC2000001625.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013363.rn.1 TC2000001625.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013371.rn.1 TC2000001625.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000013373.rn.1 TC2000001625.rn.1 RTA TC
          Rat Expression Set 230 1388307_PM_at - -
          Rat Expression Set 230 1388307_at - -
          Affymetrix Rat Gene 1.0 ST Array 10833451 - -
          Rat Genome U34 Set M13100cds#6_f_at - -
          Rat Genome U34 Set M13100cds#3_f_at - -
          Rat Genome U34 Set M13100cds#5_s_at - -
          Rat Genome U34 Set X53581cds#3_f_at - -
          Rat Genome U34 Set X05472cds#3_f_at - -
          Rat Genome U34 Set X61295cds_s_at - -
          Rat Genome U34 Set rc_AI013878_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21759451 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358918 - -
          RatRef-12 Expression BeadChip ILMN_68952 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline