Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02109864_u1
          See other SPATS2L GE Assays ›
          Gene Symbol:
          Spats2l
          Gene Name:
          spermatogenesis associated, serine-rich 2-like
          Gene Aliases:
          RGD1309930
          Chromosome Location:
          Chr.9: 64643223 - 64843963 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02109864_u1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spats2l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014102.1 NP_001014124.1 7 968 129
          XM_006244990.2 XP_006245052.1 8 840 129
          XM_006244991.2 XP_006245053.1 8 841 129
          XM_006244992.2 XP_006245054.1 9 945 129
          XM_008767108.1 XP_008765330.1 8 985 129
          XM_008767109.1 XP_008765331.1 8 798 129
          XM_008767110.1 XP_008765332.1 7 778 129
          GenBank mRNA BC085874.1 - 7 968 7191441 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000693.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spats2l

          Gene Ontology Categories:

          Process(es)

          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P504261 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P364447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393839 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150734 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87350 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P364447 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P504261 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P504261 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P364447 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P504261 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P364447 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P364447 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P504261 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61148 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29838 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80294 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3448 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60742 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65306 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10923383 - -
          Affymetrix Rat Gene 1.0 ST Array 10923384 - -
          Affymetrix Rat Gene 1.0 ST Array 10923382 - -
          Affymetrix Rat Gene 1.1 ST Array 10923381 - -
          Rat Genome 230 2.0 Array 1395336_PM_at - -
          Rat Genome 230 2.0 Array 1388791_PM_at - -
          Rat Genome 230 2.0 Array 1388791_at - -
          Rat Genome 230 2.0 Array 1395336_at - -
          Affymetrix HT Rat Focus Array 1395336_PM_at - -
          Affymetrix HT Rat Focus Array 1395336_at - -
          Affymetrix HT Rat Focus Array 1388791_PM_at - -
          Affymetrix HT Rat Focus Array 1388791_at - -
          Affymetrix Rat Clariom S Assay PSR0900005451.rn.1 TC0900000693.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005452.rn.1 TC0900000693.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005450.rn.1 TC0900000693.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005453.rn.1 TC0900000693.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005457.rn.1 TC0900000693.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005448.rn.1 TC0900000693.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005454.rn.1 TC0900000693.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005456.rn.1 TC0900000693.rn.1 RTA TC
          Rat Expression Set 230 1388791_at - -
          Rat Expression Set 230 1388791_PM_at - -
          Rat Expression Set 230 1395336_PM_at - -
          Rat Expression Set 230 1395336_at - -
          Affymetrix Rat Gene 1.0 ST Array 10923381 - -
          Rat Genome U34 Set rc_AI014140_at - -
          Rat Genome U34 Set rc_AI045262_at - -
          Rat Genome U34 Set rc_AI059272_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361905 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0