Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02111167_s1
          See other KCNJ1 GE Assays ›
          Gene Symbol:
          Kcnj1
          Gene Name:
          potassium channel, inwardly rectifying subfamily J, member 1
          Gene Aliases:
          Kcnj, ROMK1
          Chromosome Location:
          Chr.8: 33490280 - 33519116 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02111167_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnj1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017023.1 NP_058719.1 2 496 140
          XM_008766044.1 XP_008764266.1 3 627 140
          XM_008766045.1 XP_008764267.1 3 1317 140
          XM_008766046.1 XP_008764268.1 2 505 140
          XM_008766047.1 XP_008764269.1 2 618 140
          XM_008766048.1 XP_008764270.1 2 1195 140
          GenBank mRNA L29403.1 - 2 445 140
          X72341.1 - 2 496 140

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnj1

          Panther Classification:

          Molecular Function -

          ion channel transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          tissue homeostasis
          potassium ion transport
          post-embryonic development
          potassium ion import
          regulation of ion transmembrane transport
          response to potassium ion
          negative regulation of apoptotic process
          renal sodium ion absorption
          cellular response to magnesium ion
          potassium ion transmembrane transport
          cardiovascular system development
          regulation of G-protein activated inward rectifier potassium channel activity
          inward rectifier potassium channel activity
          ATP binding
          phosphatidylinositol-4,5-bisphosphate binding
          ATP-activated inward rectifier potassium channel activity
          potassium ion binding
          peptide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P470683 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P395073 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P067153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403859 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119453 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067153 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P395073 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P470683 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P395073 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P470683 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067153 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P067153 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P395073 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P470683 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P470683 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74215 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10908884 - -
          Affymetrix Rat Gene 1.0 ST Array 10908894 - -
          Affymetrix Rat Gene 1.0 ST Array 10908886 - -
          Affymetrix Rat Gene 1.0 ST Array 10908890 - -
          Affymetrix Rat Gene 1.0 ST Array 10908882 - -
          Affymetrix Rat Gene 1.0 ST Array 10908892 - -
          Affymetrix Rat Gene 1.0 ST Array 10908888 - -
          Affymetrix Rat Gene 1.0 ST Array 10908895 - -
          Affymetrix Rat Gene 1.0 ST Array 10908891 - -
          Affymetrix Rat Gene 1.0 ST Array 10908883 - -
          Affymetrix Rat Gene 1.0 ST Array 10908880 - -
          Affymetrix Rat Gene 1.0 ST Array 10908889 - -
          Affymetrix Rat Gene 1.0 ST Array 10908885 - -
          Affymetrix Rat Gene 1.0 ST Array 10908881 - -
          Affymetrix Rat Gene 1.0 ST Array 10908893 - -
          Affymetrix Rat Gene 1.1 ST Array 10908879 - -
          Rat Genome 230 2.0 Array 1369451_PM_a_at - -
          Rat Genome 230 2.0 Array 1369451_a_at - -
          Affymetrix HT Rat Focus Array 1369451_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369451_a_at - -
          Affymetrix Rat Clariom S Assay PSR0800029754.rn.1 TC0800003647.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800029755.rn.1 TC0800003647.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800029756.rn.1 TC0800003647.rn.1 RTA TC
          Rat Expression Set 230 1369451_PM_a_at - -
          Rat Expression Set 230 1369451_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10908879 - -
          Rat Genome U34 Set S78154_at - -
          Rat Genome U34 Set AF081366_s_at - -
          Rat Genome U34 Set AF081365_s_at - -
          Rat Genome U34 Set AF081365_s_at - -
          Rat Genome U34 Set AF081366_s_at - -
          Rat Genome U34 Set S78154_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22088608 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363626 - -
          RatRef-12 Expression BeadChip ILMN_53624 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline