Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02111920_s1
          See other PRDM9 GE Assays ›
          Gene Symbol:
          Prdm9
          Gene Name:
          PR domain containing 9
          Gene Aliases:
          Prdm7
          Chromosome Location:
          Chr.1: 57533602 - 57555048 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02111920_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prdm9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108903.2 NP_001102373.2 10 1203 151

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          meiotic gene conversion
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          spermatogenesis
          biological_process
          positive regulation of reciprocal meiotic recombination
          histone methylation
          peptidyl-lysine methylation
          histone lysine methylation
          positive regulation of transcription from RNA polymerase II promoter
          meiotic cell cycle
          histone H3-K4 methylation
          positive regulation of meiosis I
          molecular_function
          nucleic acid binding
          recombination hotspot binding
          histone-lysine N-methyltransferase activity
          histone methyltransferase activity (H3-K4 specific)
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P094055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663744 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71024 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P440663 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10718323 - -
          Affymetrix Rat Gene 1.0 ST Array 10718322 - -
          Affymetrix Rat Gene 1.0 ST Array 10718316 - -
          Affymetrix Rat Gene 1.0 ST Array 10718318 - -
          Affymetrix Rat Gene 1.0 ST Array 10718314 - -
          Affymetrix Rat Gene 1.0 ST Array 10718320 - -
          Affymetrix Rat Gene 1.0 ST Array 10718321 - -
          Affymetrix Rat Gene 1.0 ST Array 10718313 - -
          Affymetrix Rat Gene 1.0 ST Array 10718319 - -
          Affymetrix Rat Gene 1.0 ST Array 10718315 - -
          Affymetrix Rat Gene 1.0 ST Array 10718317 - -
          Affymetrix Rat Gene 1.1 ST Array 10718311 - -
          Affymetrix Rat Clariom S Assay PSR0100036263.rn.1 TC0100004577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036266.rn.1 TC0100004577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036268.rn.1 TC0100004577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036271.rn.1 TC0100004577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036260.rn.1 TC0100004577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036262.rn.1 TC0100004577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036269.rn.1 TC0100004577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036259.rn.1 TC0100004577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036264.rn.1 TC0100004577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036265.rn.1 TC0100004577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036272.rn.1 TC0100004577.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100036270.rn.1 TC0100004577.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10718311 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21044912 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363611 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline