Search
Search
| Gene Symbol: Fbxo48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108863.1 | NP_001102333.1 | 2 | 574 | 84 | |
| XM_006251518.2 | XP_006251580.1 | 4 | 706 | 84 | ||
| XM_008770433.1 | XP_008768655.1 | 3 | 1882 | 84 | ||
| XM_008770434.1 | XP_008768656.1 | 4 | 1244 | 84 | ||
Set Membership: |
|