Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02114053_s1
          See other TM4SF20 GE Assays ›
          Gene Symbol:
          Tm4sf20
          Gene Name:
          transmembrane 4 L six family member 20
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 88520776 - 88535002 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02114053_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tm4sf20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109618.1 NP_001103088.1 1445 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002343.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P136567 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P091168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337004 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40831 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54862 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091169 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P136567 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091169 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P136567 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P136567 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P091169 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102855 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10929446 - -
          Affymetrix Rat Gene 1.0 ST Array 10929449 - -
          Affymetrix Rat Gene 1.0 ST Array 10929447 - -
          Affymetrix Rat Gene 1.0 ST Array 10929448 - -
          Affymetrix Rat Gene 1.1 ST Array 10929445 - -
          Rat Genome 230 2.0 Array 1377928_PM_at - -
          Rat Genome 230 2.0 Array 1377928_at - -
          Affymetrix HT Rat Focus Array 1377928_PM_at - -
          Affymetrix HT Rat Focus Array 1377928_at - -
          Affymetrix Rat Clariom S Assay PSR0900019243.rn.1 TC0900002343.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019244.rn.1 TC0900002343.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019245.rn.1 TC0900002343.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019246.rn.1 TC0900002343.rn.1 RTA TC
          Rat Expression Set 230 1377928_PM_at - -
          Rat Expression Set 230 1377928_at - -
          Affymetrix Rat Gene 1.0 ST Array 10929445 - -
          Rat Genome U34 Set rc_AI013728_at - -
          Rat Genome U34 Set rc_AA819655_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22085137 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0