Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02115093_u1
          See other LMAN2 GE Assays ›
          Gene Symbol:
          Lman2
          Gene Name:
          lectin, mannose-binding 2
          Gene Aliases:
          Vip36
          Chromosome Location:
          Chr.17: 9798136 - 9815820 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02115093_u1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lman2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001115024.2 NP_001108496.1 7 876 151
          GenBank mRNA BC158808.1 - 7 876 7324877 151
          FQ215124.1 - 7 1058 151
          FQ227127.1 - 7 760 151
          FQ230263.1 - 7 874 151

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000163.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Amplicon spans exons and probe does not span exons

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          retrograde vesicle-mediated transport, Golgi to ER
          positive regulation of phagocytosis
          glycoprotein binding
          mannose binding
          carbohydrate binding
          heat shock protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1037817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320403 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16483 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146609 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1037817 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1037817 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1037817 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 589 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4760 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10794074 - -
          Affymetrix Rat Gene 1.0 ST Array 10794075 - -
          Affymetrix Rat Gene 1.0 ST Array 10794073 - -
          Affymetrix Rat Gene 1.0 ST Array 10794070 - -
          Affymetrix Rat Gene 1.0 ST Array 10794069 - -
          Affymetrix Rat Gene 1.0 ST Array 10794076 - -
          Affymetrix Rat Gene 1.0 ST Array 10794072 - -
          Affymetrix Rat Gene 1.0 ST Array 10794071 - -
          Affymetrix Rat Gene 1.1 ST Array 10794068 - -
          Rat Genome 230 2.0 Array 1373815_at - -
          Rat Genome 230 2.0 Array 1373815_PM_at - -
          Affymetrix HT Rat Focus Array 1373815_PM_at - -
          Affymetrix HT Rat Focus Array 1373815_at - -
          Affymetrix Rat Clariom S Assay PSR1700001341.rn.1 TC1700000163.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700001342.rn.1 TC1700000163.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700001345.rn.1 TC1700000163.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700001343.rn.1 TC1700000163.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700001340.rn.1 TC1700000163.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700001348.rn.1 TC1700000163.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700001347.rn.1 TC1700000163.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700001344.rn.1 TC1700000163.rn.1 RTA TC
          Rat Expression Set 230 1373815_PM_at - -
          Rat Expression Set 230 1373815_at - -
          Affymetrix Rat Gene 1.0 ST Array 10794068 - -
          Rat Genome U34 Set rc_AI230208_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366666 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0