Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02132391_s1
          See other DSPP GE Assays ›
          Gene Symbol:
          Dspp
          Gene Name:
          dentin sialophosphoprotein
          Gene Aliases:
          Dsp, RDSP2
          Chromosome Location:
          Chr.14: 6926972 - 6933079 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02132391_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dspp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012790.2 NP_036922.2 4 1444 87
          GenBank mRNA AF247187.1 - 4 1444 87
          AF250374.1 - 1444 87
          AJ403971.1 - 4 1444 87
          U63111.1 - 1 217 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dspp

          Gene Ontology Categories:

          Function(s) Process(es)

          cell differentiation
          biomineral tissue development
          odontogenesis of dentin-containing tooth
          odontogenesis
          cellular response to cell-matrix adhesion
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P165800 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221947 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152708 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11566 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P165800 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P165800 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11566 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11566 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P165800 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P165800 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11566 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79788 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10775387 - -
          Affymetrix Rat Gene 1.0 ST Array 10775386 - -
          Affymetrix Rat Gene 1.0 ST Array 10775396 - -
          Affymetrix Rat Gene 1.0 ST Array 10775384 - -
          Affymetrix Rat Gene 1.0 ST Array 10775395 - -
          Affymetrix Rat Gene 1.0 ST Array 10775392 - -
          Affymetrix Rat Gene 1.0 ST Array 10775389 - -
          Affymetrix Rat Gene 1.0 ST Array 10775394 - -
          Affymetrix Rat Gene 1.0 ST Array 10775397 - -
          Affymetrix Rat Gene 1.0 ST Array 10775388 - -
          Affymetrix Rat Gene 1.0 ST Array 10775393 - -
          Affymetrix Rat Gene 1.1 ST Array 10775383 - -
          Rat Genome 230 2.0 Array 1370734_PM_a_at - -
          Rat Genome 230 2.0 Array 1370734_a_at - -
          Rat Genome 230 2.0 Array 1368744_a_at - -
          Rat Genome 230 2.0 Array 1368744_PM_a_at - -
          Rat Genome 230 2.0 Array 1368743_PM_a_at - -
          Rat Genome 230 2.0 Array 1368743_a_at - -
          Affymetrix HT Rat Focus Array 1370734_a_at - -
          Affymetrix HT Rat Focus Array 1368744_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368744_a_at - -
          Affymetrix HT Rat Focus Array 1368743_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368743_a_at - -
          Affymetrix HT Rat Focus Array 1370734_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1400009945.rn.1 TC1400001327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400009941.rn.1 TC1400001327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400009944.rn.1 TC1400001327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400009946.rn.1 TC1400001327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400009947.rn.1 TC1400001327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400009939.rn.1 TC1400001327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400009943.rn.1 TC1400001327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400009940.rn.1 TC1400001327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400009942.rn.1 TC1400001327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400009938.rn.1 TC1400001327.rn.1 RTA TC
          Rat Expression Set 230 1370734_a_at - -
          Rat Expression Set 230 1370734_PM_a_at - -
          Rat Expression Set 230 1368743_PM_a_at - -
          Rat Expression Set 230 1368744_PM_a_at - -
          Rat Expression Set 230 1368743_a_at - -
          Rat Expression Set 230 1368744_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10775383 - -
          Rat Genome U34 Set U02074_at - -
          Rat Genome U34 Set U63111_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21343480 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651121 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline