Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053891.1 | NP_446343.1 | 1 | 1074 | 134 | |
GenBank mRNA | U50707.1 | - | 1 | 1074 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |