Search
Search
| Gene Symbol: Tlr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198769.2 | NP_942064.1 | 2 | 2367 | 109 | |
| XM_008761102.1 | XP_008759324.1 | 3 | 2732 | 109 | ||
| GenBank mRNA | AY151255.1 | - | 2 | 2367 | 109 | |
Set Membership: |
|