Search
Search
| Gene Symbol: P2ry4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031680.1 | NP_113868.1 | 2 | 1140 | 69 | |
| XM_006257093.2 | XP_006257155.1 | 4 | 1780 | 69 | ||
| XM_006257094.2 | XP_006257156.1 | 3 | 1349 | 69 | ||
| XM_006257095.2 | XP_006257157.1 | 3 | 1665 | 69 | ||
| XM_008773269.1 | XP_008771491.1 | 5 | 1498 | 69 | ||
| XM_008773270.1 | XP_008771492.1 | 5 | 1480 | 69 | ||
| GenBank mRNA | Y11433.1 | - | 2 | 1140 | 69 | |
Set Membership: |
|