Search
Search
| Gene Symbol: Gpr88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031696.1 | NP_113884.1 | 2 | 3935 | 121 | |
| XM_006233220.2 | XP_006233282.1 | 2 | 3856 | 122 | ||
| XM_006233221.2 | XP_006233283.1 | 2 | 3801 | 122 | ||
| XM_006233222.2 | XP_006233284.1 | 2 | 3798 | 122 | ||
| XM_008761471.1 | XP_008759693.1 | 3 | 4415 | 122 | ||
| GenBank mRNA | AB042407.1 | - | 2 | 3935 | 121 | |
Set Membership: |
|