Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02134195_s1
          See other SMPD3 GE Assays ›
          Gene Symbol:
          Smpd3
          Gene Name:
          sphingomyelin phosphodiesterase 3, neutral membrane
          Gene Aliases:
          cca1
          Chromosome Location:
          Chr.19: 38237965 - 38321563 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02134195_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smpd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053605.1 NP_446057.1 2 540 146
          XM_006255543.2 XP_006255605.1 3 639 146
          XM_008772541.1 XP_008770763.1 2 573 146
          XM_008772542.1 XP_008770764.1 2 1543 146
          GenBank mRNA AB000215.1 - 2 540 146

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Phosphodiesterase 1700 Inventoried Rat Phosphodiesterase Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smpd3

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          sphingolipid metabolic process
          sphingomyelin metabolic process
          multicellular organism development
          peptide hormone secretion
          sphingomyelin phosphodiesterase activity
          protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P065026 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P123462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557031 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91540 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14600 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100854 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29003 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116439 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57205 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153211 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158195 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126469 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117819 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52871 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123462 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065026 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065026 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123462 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P065026 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P123462 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12868 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700886078 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74828 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90686 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71362 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73496 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15064 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27552 - -
          Agilent Rat cDNA Microarray (G4105A) 700886078 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10810824 - -
          Affymetrix Rat Gene 1.0 ST Array 10810819 - -
          Affymetrix Rat Gene 1.0 ST Array 10810827 - -
          Affymetrix Rat Gene 1.0 ST Array 10810823 - -
          Affymetrix Rat Gene 1.0 ST Array 10810822 - -
          Affymetrix Rat Gene 1.0 ST Array 10810821 - -
          Affymetrix Rat Gene 1.0 ST Array 10810826 - -
          Affymetrix Rat Gene 1.0 ST Array 10810820 - -
          Affymetrix Rat Gene 1.0 ST Array 10810825 - -
          Affymetrix Rat Gene 1.0 ST Array 10810818 - -
          Affymetrix Rat Gene 1.1 ST Array 10810817 - -
          Rat Genome 230 2.0 Array 1368991_PM_at - -
          Rat Genome 230 2.0 Array 1368991_at - -
          Affymetrix HT Rat Focus Array 1368991_PM_at - -
          Affymetrix HT Rat Focus Array 1368991_at - -
          Affymetrix Rat Clariom S Assay PSR1900011225.rn.1 TC1900001375.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900011227.rn.1 TC1900001375.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900011231.rn.1 TC1900001375.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900011226.rn.1 TC1900001375.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900011229.rn.1 TC1900001375.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900011224.rn.1 TC1900001375.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900011228.rn.1 TC1900001375.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900011223.rn.1 TC1900001375.rn.1 RTA TC
          Rat Expression Set 230 1368991_at - -
          Rat Expression Set 230 1368991_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10810817 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20985232 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360602 - -
          RatRef-12 Expression BeadChip ILMN_58268 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline