Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02134292_s1
          See other CCR3 GE Assays ›
          Gene Symbol:
          Ccr3
          Gene Name:
          chemokine (C-C motif) receptor 3
          Gene Aliases:
          Cmkbr3
          Chromosome Location:
          Chr.8: 133028829 - 133040999 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02134292_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053958.1 NP_446410.1 3 1252 80
          XM_006244178.2 XP_006244240.1 3 2030 80
          GenBank mRNA Y13400.1 - 3 1252 80

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccr3

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of endothelial cell proliferation
          mast cell chemotaxis
          chemotaxis
          inflammatory response
          G-protein coupled receptor signaling pathway
          angioblast cell migration
          positive regulation of angiogenesis
          eosinophil chemotaxis
          chemokine-mediated signaling pathway
          ERK1 and ERK2 cascade
          chemokine receptor activity
          C-C chemokine receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P100397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501692 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59493 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P100397 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P100397 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P100397 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P100397 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87023 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102502 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10914611 - -
          Affymetrix Rat Gene 1.0 ST Array 10914610 - -
          Affymetrix Rat Gene 1.1 ST Array 10914609 - -
          Rat Genome 230 2.0 Array 1369808_PM_at - -
          Rat Genome 230 2.0 Array 1369808_at - -
          Affymetrix HT Rat Focus Array 1369808_PM_at - -
          Affymetrix HT Rat Focus Array 1369808_at - -
          Affymetrix Rat Clariom S Assay PSR0800014814.rn.1 TC0800001849.rn.1 RTA TC
          Rat Expression Set 230 1369808_at - -
          Rat Expression Set 230 1369808_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10914609 - -
          Rat Genome U34 Set Y13400_at - -
          Rat Genome U34 Set Y13400_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22002714 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357098 - -
          RatRef-12 Expression BeadChip ILMN_59482 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline