Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02134439_s1
          See other SSTR3 GE Assays ›
          Gene Symbol:
          Sstr3
          Gene Name:
          somatostatin receptor 3
          Gene Aliases:
          SSTR, Smstr28
          Chromosome Location:
          Chr.7: 119760881 - 119768082 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02134439_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sstr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133522.1 NP_598206.1 3 3749 109
          XM_006241966.2 XP_006242028.1 2 3951 109
          GenBank mRNA X63574.1 - 3 3749 109

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sstr3

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          neuropeptide signaling pathway
          chemical synaptic transmission
          spermatogenesis
          negative regulation of cell proliferation
          cerebellum development
          forebrain development
          somatostatin signaling pathway
          response to starvation
          regulation of insulin secretion
          cellular response to glucocorticoid stimulus
          cellular response to estradiol stimulus
          somatostatin receptor activity
          neuropeptide binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P718331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160649 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51309 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P718331 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P718331 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P718331 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P718331 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88897 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34300 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10905314 - -
          Affymetrix Rat Gene 1.0 ST Array 10905313 - -
          Affymetrix Rat Gene 1.0 ST Array 10905315 - -
          Affymetrix Rat Gene 1.1 ST Array 10905312 - -
          Rat Genome 230 2.0 Array 1368598_at - -
          Rat Genome 230 2.0 Array 1368598_PM_at - -
          Affymetrix HT Rat Focus Array 1368598_PM_at - -
          Affymetrix HT Rat Focus Array 1368598_at - -
          Rat Expression Set 230 1368598_at - -
          Rat Expression Set 230 1368598_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10905312 - -
          Rat Genome U34 Set X63574_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21041306 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline