Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02345797_s1
          See other ZBTB1 GE Assays ›
          Gene Symbol:
          Zbtb1
          Gene Name:
          zinc finger and BTB domain containing 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 99401775 - 99425739 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02345797_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zbtb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004444.1 NP_001004444.1 1 2975 159
          XM_006240241.2 XP_006240303.1 2 3189 159
          XM_006240243.2 XP_006240305.1 2 3196 159
          GenBank mRNA AY693992.1 - 1 2975 159

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001011.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zbtb1

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          positive regulation of T cell mediated immunity
          B cell differentiation
          positive regulation of natural killer cell differentiation
          T cell differentiation in thymus
          mRNA transcription from RNA polymerase II promoter
          positive regulation of T cell differentiation
          thymus development
          protein homooligomerization
          positive regulation of pro-T cell differentiation
          nucleic acid binding
          protein homodimerization activity
          metal ion binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P18884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262759 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138413 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18884 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18884 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P18884 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18884 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98496 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10885397 - -
          Affymetrix Rat Gene 1.0 ST Array 10885399 - -
          Affymetrix Rat Gene 1.0 ST Array 10885398 - -
          Affymetrix Rat Gene 1.1 ST Array 10885396 - -
          Rat Genome 230 2.0 Array 1384482_PM_at - -
          Rat Genome 230 2.0 Array 1393652_at - -
          Rat Genome 230 2.0 Array 1393652_PM_at - -
          Rat Genome 230 2.0 Array 1384482_at - -
          Affymetrix HT Rat Focus Array 1393652_PM_at - -
          Affymetrix HT Rat Focus Array 1393652_at - -
          Affymetrix Rat Clariom S Assay PSR0600007010.rn.1 TC0600001011.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600007011.rn.1 TC0600001011.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600007013.rn.1 TC0600001011.rn.1 RTA TC
          Rat Expression Set 230 1393652_PM_at - -
          Rat Expression Set 230 1393652_at - -
          Affymetrix Rat Gene 1.0 ST Array 10885396 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22395669 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376714 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline