Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02345830_m1
          See other CLEC4A GE Assays ›
          Gene Symbol:
          Clec4a
          Gene Name:
          C-type lectin domain family 4, member A
          Gene Aliases:
          Clecsf6, Dcir1
          Chromosome Location:
          Chr.4: 156050040 - 156067518 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02345830_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clec4a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005899.2 NP_001005899.2 1 - 2 96 132
          GenBank mRNA AY363176.1 - 1 - 2 142 132
          AY494060.1 - 1 - 2 95 132
          AY581051.1 - 1 - 2 96 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001734.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clec4a

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          innate immune response
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P031417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79837 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031417 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031417 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P031417 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80082 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 317 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10858579 - -
          Affymetrix Rat Gene 1.0 ST Array 10858575 - -
          Affymetrix Rat Gene 1.0 ST Array 10858578 - -
          Affymetrix Rat Gene 1.0 ST Array 10858576 - -
          Affymetrix Rat Gene 1.0 ST Array 10858580 - -
          Affymetrix Rat Gene 1.0 ST Array 10858574 - -
          Affymetrix Rat Gene 1.1 ST Array 10858573 - -
          Rat Genome 230 2.0 Array 1382153_at - -
          Rat Genome 230 2.0 Array 1382153_PM_at - -
          Affymetrix HT Rat Focus Array 1382153_PM_at - -
          Affymetrix HT Rat Focus Array 1382153_at - -
          Affymetrix Rat Clariom S Assay PSR0400012587.rn.1 TC0400001734.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012586.rn.1 TC0400001734.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012588.rn.1 TC0400001734.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400012589.rn.1 TC0400001734.rn.1 RTA TC
          Rat Expression Set 230 1382153_PM_at - -
          Rat Expression Set 230 1382153_at - -
          Affymetrix Rat Gene 1.0 ST Array 10858573 - -
          Rat Genome U34 Set rc_AI171821_at - -
          Rat Genome U34 Set rc_AI235315_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline